Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159731.1 | NP_001153203.1 | 3 | 423 | |
NM_009107.3 | NP_033133.1 | 4 | 879 | ||
GenBank mRNA | BC058401.1 | - | 4 | 861 | |
G49237.1 | - | 4 | 611 | ||
M84819.1 | - | 4 | 524 | ||
S62948.1 | - | 4 | 723 | ||
S62952.1 | - | 3 | 379 | ||
X66225.1 | - | 4 | 611 |