Gene Transcripts

Gene Symbol
Hltf
Entrez Gene ID
20585
Gene Name
helicase-like transcription factor
Gene Aliases
AF010600, BC057116, P113, Smarca3, Snf2l3
Chromosome Location
Chr. 3: 20057811 - 20118491 on Build GRCm38
UniGene ID
Mm.209650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009210.3 NP_033236.2 3 506
GenBank mRNA AB221636.1 - 3 329
AF010600.1 - 3 506
AF165911.1 - 3 485
AK141235.1 - 3 499
AK142961.1 - 3 463
AK149897.1 - 3 544
AK166911.1 - 3 459
BC057116.1 - 3 475 6408672
BC059240.1 - 3 475 6408672
BY726838.1 - 3 464

View Full Product Details