Gene Transcripts

Gene Symbol
Surf6
Entrez Gene ID
20935
Gene Name
surfeit gene 6
Gene Aliases
D2Wsu129e, Surf-6
Chromosome Location
Chr. 2: 26890772 - 26902813 on Build GRCm38
UniGene ID
Mm.245577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009298.3 NP_033324.1 4 433
GenBank mRNA AK077989.1 - 4 435
AK132194.1 - 4 469
BC037634.1 - 4 480 4952465
X92842.1 - 4 416

View Full Product Details