Gene Transcripts

Gene Symbol
Sntb2
Entrez Gene ID
20650
Gene Name
syntrophin, basic 2
Gene Aliases
Snt2
Chromosome Location
Chr. 8: 106935750 - 107014192 on Build GRCm38
UniGene ID
Mm.30228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009229.4 NP_033255.1 7 3427
GenBank mRNA AK010705.1 - 1 839
AK040423.1 - 7 3428
AK080059.1 - 7 3424
BC085295.1 - 1 925

View Full Product Details