Gene Transcripts

Gene Symbol
Yes1
Entrez Gene ID
22612
Gene Name
Yamaguchi sarcoma viral (v-yes) oncogene homolog 1
Gene Aliases
Yes, p61-Yes
Chromosome Location
Chr. 5: 32611171 - 32687066 on Build GRCm38
UniGene ID
Mm.4558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205132.1 NP_001192061.1 2 359
NM_001205133.1 NP_001192062.1 2 359
NM_009535.3 NP_033561.1 2 821
GenBank mRNA AK035612.1 - 2 808
AK052478.1 - 2 311
AK167420.1 - 2 496
BC010594.1 - 2 195 3989783
X67677.1 - 2 748

View Full Product Details