Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009446.2 | NP_033472.1 | 4 | 779 | |
GenBank mRNA | AK132775.1 | - | 4 | 779 | |
BC050770.1 | - | 4 | 765 | 6774775 | |
BC089547.1 | - | 4 | 765 | 6771690 | |
M13442.1 | - | 4 | 767 |