Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009531.2 | NP_033557.2 | 9 | 1176 | |
GenBank mRNA | AB071144.1 | - | 9 | 1113 | |
AK004713.1 | - | 9 | 1116 | ||
AK028595.1 | - | 9 | 1114 | ||
AK166981.1 | - | 9 | 1111 | ||
AK170975.1 | - | 9 | 1108 | ||
BC052725.1 | - | 9 | 1138 | 6812198 | |
U27398.1 | - | 9 | 1084 | ||
U40005.1 | - | 8 | 915 |