Gene Transcripts

Gene Symbol
Crisp1
Entrez Gene ID
11571
Gene Name
cysteine-rich secretory protein 1
Gene Aliases
AW324342, Aeg1, CRISP-1
Chromosome Location
Chr. 17: 40293758 - 40319246 on Build GRCm38
UniGene ID
Mm.16781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009638.3 NP_033768.3 7 595
GenBank mRNA AK009449.1 - 7 596
BC011150.1 - 7 578 4165956
L05559.1 - 7 565
M92849.1 - 7 561

View Full Product Details