Gene Transcripts

Gene Symbol
Ccr6
Entrez Gene ID
12458
Gene Name
chemokine (C-C motif) receptor 6
Gene Aliases
CC-CKR-6, CCR-6, Cmkbr6, KY411
Chromosome Location
Chr. 17: 8236043 - 8257141 on Build GRCm38
UniGene ID
Mm.8007
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190333.1 NP_001177262.1 3 512
NM_001190334.1 NP_001177263.1 4 384
NM_001190335.1 NP_001177264.1 3 293
NM_001190336.1 NP_001177265.1 3 269
NM_001190337.1 NP_001177266.1 2 178
NM_001190338.1 NP_001177267.1 2 265
NM_009835.4 NP_033965.1 6 714
GenBank mRNA AB009369.1 - 2 268
AB016031.1 - 3 343
AK089712.1 - 6 715
AK154413.1 - 5 1087
BC100756.1 - 4 380 40046797
BC100757.1 - 3 289 40046796
BC100758.1 - 3 265 40046794
BC105669.1 - 2 171 40046795

View Full Product Details