Gene Transcripts

Gene Symbol
Cst9
Entrez Gene ID
13013
Gene Name
cystatin 9
Gene Aliases
M12, cresp, testatin
Chromosome Location
Chr. 2: 148835147 - 148838737 on Build GRCm38
UniGene ID
Mm.21005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009979.1 NP_034109.1 1 162
GenBank mRNA AB017157.1 - 1 137
AK006429.1 - 1 154
BC048486.1 - 1 151 6772736
Y18243.1 - 1 162

View Full Product Details