Gene Transcripts

Gene Symbol
Cyp2a4
Entrez Gene ID
13086
Gene Name
cytochrome P450, family 2, subfamily a, polypeptide 4
Gene Aliases
AI893559, Cyp15a1, D7Ucla4
Chromosome Location
Chr. 7: 26307192 - 26315088 on Build GRCm38
UniGene ID
Mm.154643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009997.2 NP_034127.2 5 761
GenBank mRNA BC010761.1 - 5 752
BC063778.1 - 5 784 5044482

View Full Product Details