Gene Transcripts

Gene Symbol
Adam24
Entrez Gene ID
13526
Gene Name
a disintegrin and metallopeptidase domain 24 (testase 1)
Gene Aliases
Dtgn5
Chromosome Location
Chr. 8: 40675094 - 40682199 on Build GRCm38
UniGene ID
Mm.67403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010086.4 NP_034216.3 2 351
GenBank mRNA AF167402.1 - 1 339
AK029528.1 - 2 352
AK133199.1 - 2 368
BC145785.1 - 1 240 40131088

View Full Product Details