Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177937.1 | NP_001171408.1 | 8 | 3020 | |
NM_001177938.1 | NP_001171409.1 | 8 | 3017 | ||
NM_001177939.1 | NP_001171410.1 | 8 | 3011 | ||
NM_001177940.1 | NP_001171411.1 | 7 | 3002 | ||
NM_001177941.1 | NP_001171412.1 | 7 | 2987 | ||
NM_001177942.1 | NP_001171413.1 | 8 | 2984 | ||
NM_001177943.1 | NP_001171414.1 | 9 | 3176 | ||
NM_001177944.1 | NP_001171415.1 | 9 | 3159 | ||
NM_010099.2 | NP_034229.1 | 8 | 3026 | ||
GenBank mRNA | AF016628.1 | - | 8 | 3029 | |
AF016629.1 | - | 8 | 3029 | ||
AJ243657.1 | - | 8 | 3008 | ||
AJ243658.1 | - | 8 | 2960 | ||
AK137551.1 | - | 8 | 2935 | ||
BC125387.1 | - | 8 | 2917 | 40129902 | |
BC144790.1 | - | 8 | 2908 | 9053338 | |
BC144791.1 | - | 8 | 2911 | 9053339 | |
DQ168876.1 | - | 8 | 3026 | ||
DQ168877.1 | - | 8 | 3017 | ||
DQ168878.1 | - | 7 | 3002 | ||
DQ168879.1 | - | 7 | 2987 | ||
DQ168880.1 | - | 9 | 3176 | ||
DQ168881.1 | - | 9 | 3159 |