Gene Transcripts
- Gene Symbol
- RAD17
- Entrez Gene ID
- 5884
- Gene Name
- RAD17 checkpoint clamp loader component
- Gene Aliases
- CCYC, HRAD17, R24L, RAD17SP, RAD24
- Chromosome Location
- Chr. 5: 69369297 - 69414801 on Build GRCh38
- UniGene ID
- Hs.16184
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278622.1 |
NP_001265551.1
|
5, 6 |
883 |
|
NM_002873.1 |
NP_002864.1
|
4, 5 |
988 |
|
NM_133338.2 |
NP_579916.1
|
6, 7 |
884 |
|
NM_133339.2 |
NP_579917.1
|
3, 4 |
441 |
|
NM_133340.2 |
NP_579918.1
|
3, 4 |
474 |
|
NM_133341.2 |
NP_579919.1
|
3, 4 |
400 |
|
NM_133342.2 |
NP_579920.1
|
5, 6 |
861 |
|
NM_133343.1 |
NP_579921.1
|
5, 6 |
826 |
|
NM_133344.2 |
NP_579922.1
|
4, 5 |
643 |
|
XM_017009679.1 |
XP_016865168.1
|
5, 6 |
703 |
|
XM_017009680.1 |
XP_016865169.1
|
4, 5 |
757 |
|
XM_017009681.1 |
XP_016865170.1
|
4, 5 |
615 |
|
GenBank mRNA |
AF017748.1 |
- |
4, 5 |
613 |
|
AF076838.1 |
- |
4, 5 |
988 |
|
AF085736.2 |
- |
3, 4 |
441 |
|
AF098533.1 |
- |
3, 4 |
420 |
|
AF098534.1 |
- |
3, 4 |
346 |
|
AF112263.1 |
- |
3, 4 |
465 |
|
AF126424.1 |
- |
6, 7 |
883 |
|
AJ001642.1 |
- |
4, 5 |
652 |
|
AJ004977.1 |
- |
4, 5 |
648 |
|
AK292487.1 |
- |
5, 6 |
861 |
|
AL122068.1 |
- |
5, 6 |
826 |
|
BC032304.1 |
- |
3, 4 |
454 |
5170876 |
BX537441.1 |
- |
5, 6 |
883 |
|
View Full Product Details