Gene Transcripts

Gene Symbol
Gabpb1
Entrez Gene ID
14391
Gene Name
GA repeat binding protein, beta 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4Tf1B, GABPB, GABPB-1, GABPB-2, GABPB1-1, GABPB1-2, NRF2B1, NRF2B2
Chromosome Location
Chr. 2: 126628907 - 126676677 on Build GRCm38
UniGene ID
Mm.293266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271467.1 NP_001258396.1 7 935
NM_001271468.1 NP_001258397.1 6 837
NM_001271470.1 NP_001258399.1 7 1002
NM_001271492.1 NP_001258421.1 6 837
NM_010249.2 NP_034379.1 6 837
NM_207669.2 NP_997552.1 6 837
GenBank mRNA AK040722.1 - 6 838
AK052828.1 - 813
AK139710.1 - 7 1002
AK140222.1 - 7 888
AK140907.1 - 6 748
AK146373.1 - 6 701
AY282800.1 - 3 294
AY282802.1 - 4 489
AY282803.1 - 3 294
AY282804.1 - 4 376
AY282805.1 - 3 377
BC013558.1 - 6 728 3592119
CA552206.1 - 3 254
CF730595.1 - 6 690
M74516.1 - 6 732
M74517.1 - 6 731

View Full Product Details