Gene Transcripts

Gene Symbol
Fjx1
Entrez Gene ID
14221
Gene Name
four jointed box 1 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 2: 102449366 - 102451792 on Build GRCm38
UniGene ID
Mm.29730
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010218.2 NP_034348.2 1 1277
GenBank mRNA AK051097.1 - 1 1254
BC051990.1 - 1 1277 5687527

View Full Product Details