Gene Transcripts
- Gene Symbol
- Wsb1
- Entrez Gene ID
- 78889
- Gene Name
- WD repeat and SOCS box-containing 1
- Gene Aliases
- 1110056B13Rik, 2700038M07Rik
- Chromosome Location
- Chr. 11: 79239382 - 79254700 on Build GRCm38
- UniGene ID
- Mm.307022
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK200135.1 |
- |
|
47 |
|
- Gene Symbol
- Fkbp4
- Entrez Gene ID
- 14228
- Gene Name
- FK506 binding protein 4
- Gene Aliases
- 59kDa, AL022792, AW208983, FKBP-4, FKBP-52, FKBP52, FKPB52, p59
- Chromosome Location
- Chr. 6: 128430107 - 128438631 on Build GRCm38
- UniGene ID
- Mm.12758
- Species
- Mus musculus
View Full Product Details