Gene Transcripts

Gene Symbol
Gna11
Entrez Gene ID
14672
Gene Name
guanine nucleotide binding protein, alpha 11
Gene Aliases
Dsk7, E430025L19
Chromosome Location
Chr. 10: 81528732 - 81545046 on Build GRCm38
UniGene ID
Mm.260925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010301.3 NP_034431.1 3 487
GenBank mRNA AK143685.1 - 3 488
BC011169.1 - 3 486 4206878
M55411.1 - 3 424

View Full Product Details