Gene Transcripts

Gene Symbol
Gng3
Entrez Gene ID
14704
Gene Name
guanine nucleotide binding protein (G protein), gamma 3
Gene Aliases
Chromosome Location
Chr. 19: 8836929 - 8839246 on Build GRCm38
UniGene ID
Mm.329700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010316.3 NP_034446.1 3 786
GenBank mRNA AK004459.1 - 3 750
AK032646.1 - 3 734
BC029680.1 - 3 670 5365311

View Full Product Details