Gene Transcripts

Gene Symbol
Id2
Entrez Gene ID
15902
Gene Name
inhibitor of DNA binding 2
Gene Aliases
AI255428, C78922, Idb2, bHLHb26
Chromosome Location
Chr. 12: 25093799 - 25096092 on Build GRCm38
UniGene ID
Mm.34871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010496.3 NP_034626.1 1 264
GenBank mRNA AK003222.1 - 1 261
AK013239.1 - 1 259
AK013378.1 - 1 261
AK135547.1 - 1 266
AK146346.1 - 1 264
AK146606.1 - 1 267
BC006921.1 - 1 241 2655173
BC053699.1 - 1 251 6515664
M69293.1 - 1 258

View Full Product Details