Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010466.2 | NP_034596.1 | 2 | 747 | |
GenBank mRNA | AB221617.1 | - | 2 | 510 | |
AK051173.1 | - | 2 | 203 | ||
AK079948.1 | - | 2 | 336 | ||
AK080107.1 | - | 2 | 309 | ||
AK083688.1 | - | 1 | 745 | ||
X07439.1 | - | 2 | 747 | ||
X07646.1 | - | 2 | 975 |