Gene Transcripts

Gene Symbol
Klrd1
Entrez Gene ID
16643
Gene Name
killer cell lectin-like receptor, subfamily D, member 1
Gene Aliases
CD94
Chromosome Location
Chr. 6: 129588092 - 129598775 on Build GRCm38
UniGene ID
Mm.391515
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010654.3 NP_034784.1 3, 4 187
GenBank mRNA AF030311.1 - 2, 3 167
AF030312.1 - 3, 4 216
AF039025.1 - 3, 4 181
AF057714.1 - 2, 3 167
AK136548.1 - 3, 4 188
AK162976.1 - 3, 4 189
BC117112.1 - 3, 4 205 40126431
BC120854.1 - 3, 4 207 40129777

View Full Product Details