Gene Transcripts

Gene Symbol
Krtap6-2
Entrez Gene ID
16701
Gene Name
keratin associated protein 6-2
Gene Aliases
AI646275, HGTpII.4
Chromosome Location
Chr. 16: 89419323 - 89420111 on Build GRCm38
UniGene ID
Mm.3524
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010673.2 NP_034803.2 1 526
GenBank mRNA AA763314.1 - 1 21
BC116356.1 - 1 525
BC116357.1 - 1 525
BC128485.1 - 1 525
BC128486.1 - 1 525
D89902.1 - 2 538

View Full Product Details