Gene Transcripts

Gene Symbol
Ly6d
Entrez Gene ID
17068
Gene Name
lymphocyte antigen 6 complex, locus D
Gene Aliases
Ly-61, Ly61, Thb
Chromosome Location
Chr. 15: 74762056 - 74763567 on Build GRCm38
UniGene ID
Mm.878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010742.1 NP_034872.1 3 194
GenBank mRNA BC025135.1 - 3 189
X63782.1 - 3 194

View Full Product Details