Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003893.2 | NP_001003893.1 | 3 | 435 | |
NM_010767.3 | NP_034897.1 | 3 | 435 | ||
GenBank mRNA | AA238260.1 | - | 3 | 423 | |
AB009459.1 | - | 3 | 425 | ||
AI529228.1 | - | 3 | 436 | ||
AJ250369.1 | - | 3 | 424 | ||
AK050052.1 | - | 3 | 412 | ||
AK149421.1 | - | 2 | 489 | ||
BC013893.1 | - | 2 | 409 | 4211293 | |
BF233497.1 | - | 3 | 433 | ||
Y19160.1 | - | 2 | 482 | ||
Y19163.1 | - | 2 | 452 |