Gene Transcripts

Gene Symbol
Mtcp1
Entrez Gene ID
17763
Gene Name
mature T cell proliferation 1
Gene Aliases
Cmc4, P13MTCP1, p8MTCP1
Chromosome Location
Chr. X: 75404846 - 75416584 on Build GRCm38
UniGene ID
Mm.16366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294277.1 NP_001281206.1 4 397
NM_010839.6 NP_034969.1 3 721
GenBank mRNA BC138811.1 - 3 323
BC138812.1 - 3 323 8861834
Z35294.1 - 3 681

View Full Product Details