Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000680.3 | NP_000671.2 | 3 | 2228 | |
NM_033302.3 | NP_150645.2 | 2 | 1920 | ||
NM_033303.4 | NP_150646.3 | 2 | 1920 | ||
NM_033304.3 | NP_150647.2 | 2 | 1920 | ||
XM_006716292.3 | XP_006716355.1 | 3 | 2255 | ||
XM_006716293.3 | XP_006716356.1 | 3 | 2255 | ||
XM_017013094.1 | XP_016868583.1 | 4 | 5815 | ||
XM_017013095.1 | XP_016868584.1 | 4 | 5838 | ||
GenBank mRNA | AF013261.1 | - | 2 | 1434 | |
AF395806.1 | - | 2 | 1239 | ||
AK289548.1 | - | 3 | 2210 | ||
BC095512.1 | - | 2 | 1246 | 6971913 | |
D25235.1 | - | 2 | 1670 | ||
D32201.1 | - | 2 | 1670 | ||
D32202.1 | - | 2 | 1670 | ||
L31774.1 | - | 2 | 1234 | ||
U02569.1 | - | 2 | 1658 |