Gene Transcripts

Gene Symbol
Reg3b
Entrez Gene ID
18489
Gene Name
regenerating islet-derived 3 beta
Gene Aliases
HIP, PAP1, Pap, REG-III
Chromosome Location
Chr. 6: 78370885 - 78373466 on Build GRCm38
UniGene ID
Mm.2553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011036.1 NP_035166.1 2, 3 108
GenBank mRNA BC027525.1 - 1, 2 91
D13509.1 - 81
D63359.1 - 2, 3 108

View Full Product Details