Gene Transcripts

Gene Symbol
Phf2
Entrez Gene ID
18676
Gene Name
PHD finger protein 2
Gene Aliases
GRC5
Chromosome Location
Chr. 13: 48801750 - 48870885 on Build GRCm38
UniGene ID
Mm.486213
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011078.3 NP_035208.2 5 598
GenBank mRNA AB221591.1 - 5 482
AF043726.1 - 5 598
AK172995.1 - 3 305
BC051633.1 - 5 596 6518591

View Full Product Details