Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007050.5 | NP_008981.4 | 31 | 12466 | |
NM_133170.3 | NP_573400.3 | 32 | 12523 | ||
XM_017027611.1 | XP_016883100.1 | 31 | 12559 | ||
XM_017027612.1 | XP_016883101.1 | 31 | 12556 | ||
XM_017027613.1 | XP_016883102.1 | 30 | 12498 | ||
GenBank mRNA | AB006621.2 | - | 31 | 12374 | |
AF043644.4 | - | 32 | 12531 | ||
BC153300.1 | - | 31 | 12374 | 8860135 |