Gene Transcripts

Gene Symbol
Psmb5
Entrez Gene ID
19173
Gene Name
proteasome (prosome, macropain) subunit, beta type 5
Gene Aliases
Chromosome Location
Chr. 14: 54614120 - 54617995 on Build GRCm38
UniGene ID
Mm.8911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011186.1 NP_035316.1 2 256
GenBank mRNA AB003304.1 - 2 259
AF060091.1 - 2 256
AK010441.1 - 2 276
AK133839.1 - 2 277
AK219104.1 - 2 84
BC012246.1 - 2 264 4167355
BC106144.1 - 2 259 4953887

View Full Product Details