Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011192.3 | NP_035322.1 | 5 | 467 | |
GenBank mRNA | AK140597.1 | - | 5 | 488 | |
AK150939.1 | - | 5 | 487 | ||
AK156079.1 | - | 5 | 437 | ||
AK171348.1 | - | 5 | 483 | ||
AK182764.1 | - | 3 | 211 | ||
AK183517.1 | - | 3 | 211 | ||
AK183726.1 | - | 3 | 172 | ||
AK184446.1 | - | 3 | 211 | ||
AK198462.1 | - | 3 | 211 | ||
AK210357.1 | - | 3 | 207 | ||
BC015255.1 | - | 5 | 467 | 4221225 | |
BC139818.1 | - | 5 | 415 | 40141268 | |
CT010198.1 | - | 5 | 271 | ||
D87911.1 | - | 5 | 390 | ||
U60330.1 | - | 5 | 271 |