Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152430.3 | NP_689643.2 | 2 | 1049 | |
GenBank mRNA | AL833127.1 | - | 2 | 994 | |
AY698056.1 | - | 2 | 994 | ||
AY775731.1 | - | 1 | 1102 | ||
BC022401.1 | - | 2 | 1049 | 4694856 |