Gene Transcripts

Gene Symbol
Scn8a
Entrez Gene ID
20273
Gene Name
sodium channel, voltage-gated, type VIII, alpha
Gene Aliases
AI853486, C630029C19Rik, NaCh6, Nav1.6, dmu, med, mnd-2, mnd2, nmf2, nmf335, nmf58, nur14, seal
Chromosome Location
Chr. 15: 100868549 - 101045938 on Build GRCm38
UniGene ID
Mm.385012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077499.2 NP_001070967.1 2 407
NM_011323.3 NP_035453.2 1 308
GenBank mRNA AF049617.1 - 1 254
AK158609.1 - 2 369
BC099466.1 - 2 375
CJ128345.1 - 2 408
U26707.1 - 1 266

View Full Product Details