Gene Transcripts

Gene Symbol
Itsn2
Entrez Gene ID
20403
Gene Name
intersectin 2
Gene Aliases
AI327390, Ese2, Sh3d1B, Sh3p18, mKIAA1256
Chromosome Location
Chr. 12: 4592638 - 4713952 on Build GRCm38
UniGene ID
Mm.341204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198968.2 NP_001185897.1 20, 21 2584
NM_011365.4 NP_035495.2 19, 20 2503
GenBank mRNA AF132479.1 - 18, 19 2243
AF132480.1 - 18, 19 2243
AK122480.1 - 14, 15 1890
AK161743.1 - 8, 9 921
BC138262.1 - 20, 21 2561 8861281
BC171950.1 - 20, 21 2561 9054236
BI872589.1 - 4, 5 471

View Full Product Details