Gene Transcripts

Gene Symbol
Plk4
Entrez Gene ID
20873
Gene Name
polo-like kinase 4
Gene Aliases
1700028H20, AI385771, Sak, Stk18
Chromosome Location
Chr. 3: 40799951 - 40816883 on Build GRCm38
UniGene ID
Mm.3794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011495.2 NP_035625.2 4, 5 643
NM_173169.2 NP_775261.2 4, 5 643
GenBank mRNA AK006827.1 - 4, 5 549
AK045082.1 - 4, 5 567
AK137080.1 - 4, 5 555
AK137471.1 - 4, 5 554
BC026785.1 - 4, 5 553 5150173
BC057940.1 - 4, 5 540 5323295
L29479.1 - 4, 5 545
L29480.1 - 4, 5 545

View Full Product Details