Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079690.1 | NP_001073158.1 | 13 | 1800 | |
NM_183354.2 | NP_899197.2 | 13 | 1800 | ||
GenBank mRNA | BC016888.1 | - | 12 | 1778 | |
BC051100.1 | - | 6 | 564 | ||
U20973.1 | - | 13 | 1798 | ||
U20974.1 | - | 13 | 1798 | ||
U20975.1 | - | 13 | 1798 | ||
U61381.1 | - | 12 | 1769 | ||
U94518.1 | - | 12 | 1747 |