Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011465.4 | NP_035595.2 | 44 | 6452 | |
GenBank mRNA | AF093576.1 | - | 44 | 6360 | |
AK086017.1 | - | 1 | 92 | ||
AK143823.1 | - | 8 | 1120 | ||
AK161449.1 | - | 14 | 3378 | ||
BC150747.1 | - | 44 | 6317 | 9087658 | |
U87455.1 | - | 44 | 6432 |