Gene Transcripts

Gene Symbol
Tekt1
Entrez Gene ID
21689
Gene Name
tektin 1
Gene Aliases
MT14
Chromosome Location
Chr. 11: 72344717 - 72362442 on Build GRCm38
UniGene ID
Mm.42257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282006.1 NP_001268935.1 3 370
NM_001282007.1 NP_001268936.1 3 319
NM_011569.3 NP_035699.2 3 322
GenBank mRNA AF081947.1 - 3 313
AK005801.1 - 3 371
AK076858.1 - 3 300
BC050967.1 - 3 320
BC098181.1 - 3 280 6312556
BY085053.1 - 3 321

View Full Product Details