Gene Transcripts

Gene Symbol
Myo18a
Entrez Gene ID
360013
Gene Name
myosin XVIIIA
Gene Aliases
MAJN, MyoPDZ, MysPDZ
Chromosome Location
Chr. 11: 77763251 - 77865988 on Build GRCm38
UniGene ID
Mm.341248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291212.1 NP_001278141.1 14, 15 2627
NM_001291213.1 NP_001278142.1 14, 15 2735
NM_001291214.1 NP_001278143.1 12, 13 1582
NM_001291215.1 NP_001278144.1 15, 16 1648
NM_011586.3 NP_035716.1 13, 14 2591
GenBank mRNA AB026497.1 - 13, 14 2561
AK137574.1 - 13, 14 1582
AK147584.1 - 15, 16 1597
AK171342.1 - 12, 13 1579
BC138366.1 - 14, 15 2569 8861386
BC145257.1 - 14, 15 2677

View Full Product Details