Gene Transcripts

Gene Symbol
Cfdp1
Entrez Gene ID
23837
Gene Name
craniofacial development protein 1
Gene Aliases
AA408409, Bcnt, Bucentaur, Cfdp, cp27
Chromosome Location
Chr. 8: 111768473 - 111854310 on Build GRCm38
UniGene ID
Mm.279437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011801.1 NP_035931.1 2 243
GenBank mRNA AB010828.1 - 2 243
AK133989.1 - 2 261
AK160532.1 - 2 208
BC005589.1 - 2 224 3964401
Y08219.1 - 2 242

View Full Product Details