Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159500.1 | NP_001152972.1 | 2 | 674 | |
NM_011934.4 | NP_036064.3 | 2 | 573 | ||
GenBank mRNA | AK032025.1 | - | 2 | 381 | |
AK044339.1 | - | 2 | 574 | ||
AK052256.1 | - | 2 | 676 | ||
BC044858.1 | - | 1 | 243 | ||
BC132595.1 | - | 2 | 538 | 40130872 | |
BC132597.1 | - | 2 | 539 | 40130874 | |
S82458.1 | - | 1 | 281 | ||
X89594.1 | - | 1 | 281 |