Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011937.2 | NP_036067.2 | 2 | 190 | |
GenBank mRNA | AF088903.1 | - | 2 | 148 | |
AF160355.1 | - | 2 | 136 | ||
AK080610.1 | - | 2 | 138 | ||
AK084247.1 | - | 1 | 820 | ||
AK166069.1 | - | 2 | 140 | ||
AK172879.1 | - | 1 | 53 | ||
AK200561.1 | - | 1 | 50 | ||
BC014800.1 | - | 2 | 121 | 4220980 | |
BY092839.1 | - | 2 | 165 | ||
CJ048539.1 | - | 2 | 191 |