Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011933.2 | NP_036063.1 | 4 | 405 | |
GenBank mRNA | AF155575.1 | - | 4 | 405 | |
AK137212.1 | - | 4 | 336 | ||
AK149270.1 | - | 4 | 530 | ||
AK162961.1 | - | 4 | 541 | ||
BC021865.1 | - | 4 | 377 | 5123899 |