Gene Transcripts
- Gene Symbol
- Orc4
- Entrez Gene ID
- 26428
- Gene Name
- origin recognition complex, subunit 4
- Gene Aliases
- Orc4P, Orc4l, mMmORC4
- Chromosome Location
- Chr. 2: 48902754 - 48950272 on Build GRCm38
- UniGene ID
- Mm.291059
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177313.1 |
NP_001170784.1
|
4, 5 |
384 |
|
NM_011958.3 |
NP_036088.3
|
4, 5 |
384 |
|
GenBank mRNA |
AJ003140.1 |
- |
4, 5 |
345 |
|
AK010258.1 |
- |
4, 5 |
368 |
|
AK011550.1 |
- |
4, 5 |
350 |
|
AK012669.1 |
- |
4, 5 |
352 |
|
AK049534.1 |
- |
4, 5 |
352 |
|
AK076324.1 |
- |
4, 5 |
385 |
|
AK084326.1 |
- |
4, 5 |
353 |
|
AK145454.1 |
- |
4, 5 |
395 |
|
AK169291.1 |
- |
4, 5 |
368 |
|
BC015072.1 |
- |
4, 5 |
356 |
4212266 |
BC019748.1 |
- |
4, 5 |
326 |
5007346 |
Y16386.1 |
- |
3, 4 |
216 |
|
View Full Product Details