Gene Transcripts

Gene Symbol
Actn3
Entrez Gene ID
11474
Gene Name
actinin alpha 3
Gene Aliases
Chromosome Location
Chr. 19: 4861216 - 4877909 on Build GRCm38
UniGene ID
Mm.5316
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013456.2 NP_038484.1 20, 21 2599
GenBank mRNA AF093775.1 - 20, 21 2574
AK134757.1 - 5, 6 703
BC111890.1 - 20, 21 2565 40060598

View Full Product Details