Gene Transcripts

Gene Symbol
Drd5
Entrez Gene ID
13492
Gene Name
dopamine receptor D5
Gene Aliases
D5R, Drd-5, Drd1b, Gpcr1
Chromosome Location
Chr. 5: 38319509 - 38322310 on Build GRCm38
UniGene ID
Mm.167154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK032054.1 - 1 951
AK045456.1 - 1 1140
BC138059.1 - 1 1008 8861078
BC138060.1 - 1 1008 8861079

View Full Product Details