Gene Transcripts

Gene Symbol
Ksr1
Entrez Gene ID
16706
Gene Name
kinase suppressor of ras 1
Gene Aliases
AW492498, B-KSR1, D11Bhm183e, D11Bhm184e, Ksr, RSU2, hb
Chromosome Location
Chr. 11: 79013440 - 79146554 on Build GRCm38
UniGene ID
Mm.4745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013571.2 NP_038599.1 1, 2 308
GenBank mRNA AK036566.1 - 1, 2 364
AK164833.1 - 1, 2 428
U43585.1 - 1, 2 315
X81634.1 - 1, 2 90

View Full Product Details