Gene Transcripts
- Gene Symbol
- Lifr
- Entrez Gene ID
- 16880
- Gene Name
- leukemia inhibitory factor receptor
- Gene Aliases
- A230075M04Rik, AW061234, LIF
- Chromosome Location
- Chr. 15: 7090611 - 7197489 on Build GRCm38
- UniGene ID
- Mm.149720
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113386.1 |
NP_001106857.1
|
4, 5 |
674 |
|
NM_013584.2 |
NP_038612.1
|
3, 4 |
323 |
|
GenBank mRNA |
AK145864.1 |
- |
4, 5 |
675 |
|
AK155524.1 |
- |
3, 4 |
550 |
|
BC031929.1 |
- |
3, 4 |
316 |
4159053 |
BY716975.1 |
- |
3, 4 |
541 |
|
D17444.1 |
- |
3, 4 |
330 |
|
D26177.1 |
- |
3, 4 |
330 |
|
S73495.1 |
- |
3, 4 |
330 |
|
S73496.1 |
- |
3, 4 |
330 |
|
View Full Product Details