Gene Transcripts

Gene Symbol
Prl3c1
Entrez Gene ID
27372
Gene Name
prolactin family 3, subfamily c, member 1
Gene Aliases
PLP-J, Prlpi, Prlpj, decidualin
Chromosome Location
Chr. 13: 27196660 - 27203749 on Build GRCm38
UniGene ID
Mm.291456
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163218.1 NP_001156690.1 4 510
NM_013766.2 NP_038794.1 4 480
GenBank mRNA AB019118.1 - 4 477
AF150740.1 - 4 490
AF234637.1 - 4 480
AK014980.1 - 4 510
BC120576.1 - 4 468 40129499
BC120578.1 - 4 468 40129501

View Full Product Details